Chromium (III) sulphate hydrate, Reagent Grade
Leverantör: Thermo Fisher Scientific
Varning
33303.22 33303.36
33303.22EA 2150 SEK
33303.22 33303.36
Chromium (III) sulphate hydrate, Reagent Grade
Chromium (III) sulphate hydrate
CAS No.: 15244-38-9 Molecular Formula: Cr2(SO4)3.xH2O Formula Weight: 392.17(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Green to violet Odour: Odourless MDL No.: MFCD00016039
Formel: Cr₂O₁₂S₃·H₂O
MW: 392.17 g/mol (anhy)
Storage Temperature: Omgivande
MDL Number: MFCD00016039
CAS nummer: 15244-38-9
EINECS: 233-253-2