Sodium bis(2-methoxyethoxy)aluminium hydride ≥70% (w/w) I toluen
Leverantör: Thermo Scientific
Fara
Synonyms: Sodium dihydrobis(2-methoxyethoxy)aluminate, SBAH, Sodium bis(2-methoxyethoxy)aluminum dihydride
A13292.0B A13292.18 A13292.30
A13292.0BEA 5950 SEK
A13292.0B A13292.18 A13292.30
Sodium bis(2-methoxyethoxy)aluminium hydride ≥70% (w/w) I toluen
Sodium bis(2-methoxyethoxy)aluminium hydride
CAS No.: 22722-98-1 Molecular Formula: C6H16AlNaO4 Formula Weight: 202.16 Flash Point: 4°(39°F) Merck Reference: 14,8565 UN No.: 3399 MDL No.: MFCD00011631
Formel: C₆H₁₆AlNaO₄
MW: 202,16 g/mol
Densitet: 1,020
Flampunkt: 4 °C (39 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00011631
CAS nummer: 22722-98-1
EINECS: 245-178-2
UN: 3399
ADR: 4.3,I
Merck Index: 14,8565