Gadolinium hexaboride ≥99.5%
Leverantör: Thermo Scientific
Synonyms:
Gadolinium boride
39225.03
39225.06
39225.03EA
1150
SEK
39225.03
39225.06
Gadolinium hexaboride ≥99.5%
Gadolinium hexaboride
Purity: 99.5%
CAS No.: 12008-06-9
Molecular Formula: GdB6
Formula Weight: 222.12
Physical Form: -325 Mesh Powder
Odour: Odourless
MDL No.: MFCD00049942
Formel:
B₆Gd MW: 222,12 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00049942 CAS nummer: 12008-06-9 EINECS: 234-530-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...