2,3-Dichloroanisole ≥97.0%

Leverantör: TCI EUROPE

Synonyms: 1,2-Dichloro-3-methoxybenzene

D2306-25G D2306-500G
TCIAD2306-25GEA 535 SEK
TCIAD2306-25G TCIAD2306-500G
2,3-Dichloroanisole ≥97.0%
2,3-Dichloroanisole
Formel: C₇H₆Cl₂O
MW: 177,03 g/mol
Kokpunkt: 128 °C (20 -rr)
Smältpunkt: 31…34 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000545
CAS nummer: 1984-59-4
EINECS: 217-853-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a89d372e9ceed4a7b7909da95844b46463447b65&vid=b64ca5a0b302d51e7e7fa28e43ed27cfc747bc03&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to lump
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR