Nessler reagent
Leverantör: Lovibond
Fara
Synonyms: Potassium mercuric Iodide, Potassium tetraiodomercurate, Nota 1, Mercury (II) potassium iodide, Potassium tetraiodomercurate – Potassium hydroxide solution
465200 465201
LOVI465200EA 330 SEK
LOVI465200 LOVI465201
Nessler reagent
Nessler reagent
MW: 786,4 g/mol
Kokpunkt: ∼100 °C (1013 hPa)
Densitet: 1,2 g/cm³ (20 °C)
MDL Number: MFCD00049656
CAS nummer: 7783-33-7
UN: 2922
ADR: 8,II