2-Bromo-5-iodotoluene ≥98.0%
Leverantör: TCI
|
Varning
|
B3099-25G
B3099-5G
TCIAB3099-25GEA
2875
SEK
TCIAB3099-25G
TCIAB3099-5G
2-Bromo-5-iodotoluene ≥98.0%
2-Bromo-5-iodotoluene
Formel:
C₇H₆BrI MW: 296.93 g/mol |
MDL Number:
MFCD00070749 CAS nummer: 202865-85-8 |
|
Specifikation testresultat
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...