Propanephosphonic acid cyclic anhydride ≥50% i DMF
Leverantör: Thermo Scientific
Fara
Synonyms: 1-Propylphosphonic anhydride, 2,4,6-Tripropyl-1,3,5,2,4,6-trioxotriphosphorinane-2,4,6-trioxide
L19271.09 L19271.18
L19271.09EA 785 SEK
L19271.09 L19271.18
Propanephosphonic acid cyclic anhydride ≥50% i DMF
Propanephosphonic acid cyclic anhydride
CAS No.: 68957-94-8 Molecular Formula: C9H21O6P3 Formula Weight: 318.19 Flash Point: 62°(143°F) Refractive Index: 1.4505 Boiling Point: 153° UN No.: 3265 MDL No.: MFCD00006583
Formel: C₉H₂₁O₆P₃
MW: 318,19 g/mol
Kokpunkt: 153 °C
Densitet: 1,090
Flampunkt: 62 °C (143 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00006583
CAS nummer: 68957-94-8
EINECS: 422-210-5
UN: 3265
ADR: 8,II