Propanephosphonic acid cyclic anhydride ≥50% i etylacetat
Leverantör: Thermo Scientific
Fara
Synonyms: 1-Propylphosphonic anhydride, 2,4,6-Tripropyl-1,3,5,2,4,6-trioxotriphosphorinane-2,4,6-trioxide
L11911.09 L11911.18
L11911.09EA 1150 SEK
L11911.09 L11911.18
Propanephosphonic acid cyclic anhydride ≥50% i etylacetat
Propanephosphonic acid cyclic anhydride
CAS No.: 68957-94-8 Molecular Formula: C9H21O6P3 Formula Weight: 318.19 Flash Point: -3°(26°F) Refractive Index: 1.4190 Boiling Point: 77° UN No.: 2924 MDL No.: MFCD00006583
Formel: C₉H₂₁O₆P₃
MW: 318,19 g/mol
Kokpunkt: 77 °C
Densitet: 1,080
Flampunkt: –3 °C (26 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00006583
CAS nummer: 68957-94-8
EINECS: 422-210-5
UN: 2924
ADR: 3,II