1,3,2-Benzodioxaborole (catecholborane) 50% (w/w) I toluen , 98%
Leverantör: Thermo Scientific
Fara
H30234.14 H30234.22
H30234.14EA 7350 SEK
H30234.14 H30234.22
1,3,2-Benzodioxaborole (catecholborane) 50% (w/w) I toluen , 98%
1,3,2-Benzodioxaborole (catecholborane)
CAS No.: 274-07-7 Molecular Formula: C6H5BO2 Formula Weight: 119.92 Refractive Index: 1.5025 UN No.: 2924 MDL No.: MFCD00005846
Formel: C₆H₅BO₂
MW: 119,92 g/mol
Densitet: 0,911
Storage Temperature: Omgivande
MDL Number: MFCD00005846
CAS nummer: 274-07-7
EINECS: 205-991-5
UN: 2924
ADR: 3,II