Isophorondiamine cis- and trans-mixture 99+%
Leverantör: Thermo Fisher Scientific
Fara
352942500 352940010
ACRO352942500EA 1475 SEK
ACRO352942500 ACRO352940010
Isophorondiamine cis- and trans-mixture 99+%
Isophorondiamine
Formel: C₁₀H₂₂N₂
Kokpunkt: 247 °C
Smältpunkt: 10 °C
Flampunkt: 117 °C
Storage Temperature: Omgivande
MDL Number: MFCD00019397
CAS nummer: 2855-13-2
EINECS: 220-666-8
UN: 2289
ADR: 8,III
Order Now
Specifikation testresultat
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4880 to 1.4900 (20°C, 589 nm)
GC ≥99.0 %
Color scale ≤15 APHA