2,6-Pyridinedimethanol ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 2,6-Bis(hydroxymethyl)pyridine
L02527.06 L02527.14
L02527.06EA 1750 SEK
L02527.06 L02527.14
2,6-Pyridinedimethanol ≥98%
2,6-Pyridinedimethanol
Purity: 98% CAS No.: 1195-59-1 Molecular Formula: C7H9NO2 Formula Weight: 139.15 Melting Point: 110-114° Boiling Point: 185°/15mm MDL No.: MFCD00006351
Formel: C₇H₉NO₂
MW: 139.15 g/mol
Kokpunkt: 185 °C (15 mmHg)
Smältpunkt: 110…114 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006351
CAS nummer: 1195-59-1
EINECS: 214-803-0