Naphthalene-1,4-dicarbonitrile 98+%
Leverantör: Thermo Scientific
Varning
L20248.06 L20248.14
L20248.06EA 2300 SEK
L20248.06 L20248.14
Naphthalene-1,4-dicarbonitrile 98+%
Naphthalene-1,4-dicarbonitrile
Purity: 98+% CAS No.: 3029-30-9 Molecular Formula: C12H6N2 Formula Weight: 178.19 Odour: Odourless Melting Point: 208-212° Beilstein Reference: 1870226 MDL No.: MFCD03093626
Formel: C₁₂H₆N₂
MW: 178,19 g/mol
Smältpunkt: 208…212 °C
Storage Temperature: Omgivande
MDL Number: MFCD03093626
CAS nummer: 3029-30-9
EINECS: 221-197-1