Phenyl phosphorodiamidate 98+%, rods
Leverantör: Thermo Fisher Scientific
|
Fara
|
A13562.09
A13562.18
A13562.09EA
7875
SEK
A13562.09
A13562.18
Phenyl phosphorodiamidate 98+%, rods
Phenyl phosphorodiamidate
Purity: 98+%
CAS No.: 7450-69-3
Molecular Formula: C6H9N2O2P
Formula Weight: 172.12
Melting Point: 180-185°
Beilstein Reference: 608629
Formel:
C₆H₉N₂O₂P MW: 172.12 g/mol Smältpunkt: 180…185 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00014767 CAS nummer: 7450-69-3 EINECS: 231-218-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...