Diphenyl phosphoramidate ≥97%
Leverantör: Thermo Fisher Scientific
L07155.06
L07155.14
L07155.06EA
1475
SEK
L07155.06
L07155.14
Diphenyl phosphoramidate ≥97%
Diphenyl phosphoramidate
Purity: 97%
CAS No.: 2015-56-7
Molecular Formula: C12H12NO3P
Formula Weight: 249.21
Melting Point: 147-151°
Beilstein Reference: 2054166
Formel:
C₁₂H₁₂NO₃P MW: 249.21 g/mol |
MDL Number:
MFCD00014078 CAS nummer: 2015-56-7 |