4,4'-Dimethoxytrityl chloride ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: DMT-Cl, 4,4'-Dimethoxytriphenylmethyl chloride
A11626.06 A11626.14 A11626.22
A11626.06EA 1150 SEK
A11626.06 A11626.14 A11626.22
4,4'-Dimethoxytrityl chloride ≥98%
4,4'-Dimethoxytrityl chloride
Purity: 98% CAS No.: 40615-36-9 Molecular Formula: C21H19ClO2 Formula Weight: 338.83 Melting Point: 121-127° Beilstein Reference: 2471942
Formel: C₂₁H₁₉ClO₂
MW: 338,8 g/mol
Smältpunkt: 121…127 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008409
CAS nummer: 40615-36-9
EINECS: 255-002-6