Iminodiacetic acid ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 2,2'-Azanediyldiacetic acid
A11510.0E A11510.22 A11510.36
A11510.0EEA 5325 SEK
A11510.0E A11510.22 A11510.36
Iminodiacetic acid ≥98%
Iminodiacetic acid
Purity: 98+% CAS No.: 142-73-4 Molecular Formula: HN(CH2CO2H)2 Formula Weight: 133.10 Melting Point: ca 243° dec. Beilstein Reference: 878499 Merck Reference: 14,4917 MDL No.: MFCD00004280
Formel: C₄H₇NO₄
MW: 133,10 g/mol
Smältpunkt: ∼ 243 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00004280
CAS nummer: 142-73-4
EINECS: 205-555-4
Merck Index: 14,4917