1-Methylimidazole-5-carboxylic acid

Leverantör: Thermo Scientific

Synonyms: 1-Methylimidazole-5-carboxylic Acid

CC23601DA
MAYBCC23601DAEA 0 SEK
MAYBCC23601DA
1-Methylimidazole-5-carboxylic acid
1-Methylimidazole-5-carboxylic acid
Formel: C₅H₆N₂O₂
MW: 126,1133 g/mol
MDL Number: MFCD00955677
CAS nummer: 41806-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR