1-Methylimidazole-2-carboxylic acid

Leverantör: Thermo Scientific

Synonyms: 1-Methyl-1H-imidazole-2-carboxylic acid

CC20801DA
MAYBCC20801DAEA 0 SEK
MAYBCC20801DA
1-Methylimidazole-2-carboxylic acid
1-Methylimidazole-2-carboxylic acid
Formel: C₅H₆N₂O₂
MW: 126,11 g/mol
MDL Number: MFCD01863421
CAS nummer: 20485-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR