1-Methylimidazole-4-carboxylic acid methyl ester

Leverantör: Thermo Scientific

Synonyms: Methyl 1-methyl-1H-imidazole-4-carboxylate

CC03622DA CC03622EA
MAYBCC03622DAEA 0 SEK
MAYBCC03622DA MAYBCC03622EA
1-Methylimidazole-4-carboxylic acid methyl ester
1-Methylimidazole-4-carboxylic acid methyl ester
Formel: C₆H₈N₂O₂
MW: 140.1400 g/mol
MDL Number: MFCD02179561
CAS nummer: 17289-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR