2,4-Hexadiyne-1,6-diol ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: Hexa-2,4-diyne-1,6-diol
L09576.03 L09576.06
L09576.03EA 1075 SEK
L09576.03 L09576.06
2,4-Hexadiyne-1,6-diol ≥97%
2,4-Hexadiyne-1,6-diol
Purity: 97% CAS No.: 3031-68-3 Molecular Formula: C6H6O2 Formula Weight: 110.11 Melting Point: 110-114° Beilstein Reference: 773791 UN No.: 1325 MDL No.: MFCD00020619
Formel: C₆H₆O₂
MW: 110,11 g/mol
Smältpunkt: 110…114 °C
Storage Temperature: Omgivande
MDL Number: MFCD00020619
CAS nummer: 3031-68-3
EINECS: 221-210-0
UN: 1325
ADR: 4.1,III