Diethyl (iodomethyl)phosphonate ≥98%
Leverantör: Thermo Fisher Scientific
Varning
A13697.06 A13697.14 A13697.22
A13697.06EA 1875 SEK
A13697.06 A13697.14 A13697.22
Diethyl (iodomethyl)phosphonate ≥98%
Diethyl (iodomethyl)phosphonate
Purity: 98+% CAS No.: 10419-77-9 Molecular Formula: C5H12IO3P Formula Weight: 278.03 Flash Point: 110°(230°F) Refractive Index: 1.4995 Boiling Point: 112-114°/1mm Beilstein Reference: 1812052 MDL No.: MFCD00041386
Formel: C₅H₁₂IO₃P
Kokpunkt: 112…114 °C (1 mmHg)
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00041386
CAS nummer: 10419-77-9
EINECS: 233-895-3