Triethyl-2-phosphonobutyrate 98%
Leverantör: Thermo Scientific
Varning
Synonyms: Ethyl 2-(diethoxyphosphoryl)butanoate
368780050 368780250
ACRO368780050EA 710 SEK
ACRO368780050 ACRO368780250
Triethyl-2-phosphonobutyrate 98%
Triethyl-2-phosphonobutyrate
Formel: C₁₀H₂₁O₅P
MW: 252,25 g/mol
Kokpunkt: 152…154 °C (14 mmHg)
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00041347
CAS nummer: 17145-91-4
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4315 to 1.4335 (20°C, 589 nm)
GC ≥97.5 %