N,N-Diethylcarbamoyl chloride ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Diethylcarbamyl chloride
A15701.22 A15701.14
A15701.22EA 1500 SEK
A15701.22 A15701.14
N,N-Diethylcarbamoyl chloride ≥98%
N,N-Diethylcarbamoyl chloride
Purity: 98% CAS No.: 88-10-8 Molecular Formula: C5H10ClNO Formula Weight: 135.59 Flash Point: 76°(168°F) Refractive Index: 1.4510 Melting Point: -32° Boiling Point: 186° Beilstein Reference: 506687 UN No.: 3265 MDL No.: MFCD00000636
Formel: C₅H₁₀ClNO
MW: 135,59 g/mol
Kokpunkt: 186 °C
Smältpunkt: –32 °C
Densitet: 1,070
Flampunkt: 76 °C (168 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000636
CAS nummer: 88-10-8
EINECS: 201-798-5
UN: 3265
ADR: 8,II