2,4-Dichloro-5-methoxypyrimidine ≥98.0%

Leverantör: TCI EUROPE
Varning

D3508-25G D3508-5G
TCIAD3508-25GEA 1950 SEK
TCIAD3508-25G TCIAD3508-5G
2,4-Dichloro-5-methoxypyrimidine ≥98.0%
2,4-Dichloro-5-methoxypyrimidine
Formel: C₅H₄Cl₂N₂O
MW: 179,00 g/mol
MDL Number: MFCD02685670
CAS nummer: 19646-07-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=abfbcc51c79ca597cfa93f31cba7e284783e72d2&vid=abfbcc51c79ca597cfa93f31cba7e284783e72d2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 66.0 to 72.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR