Methyl-4-chloroacetoacetate 97%

Leverantör: Thermo Scientific
Fara

Synonyms: Methyl 4-chloro-3-oxobutanoate

241620250 241621000
ACRO241620250EA 1375 SEK
ACRO241620250 ACRO241621000
Methyl-4-chloroacetoacetate 97%
Methyl-4-chloroacetoacetate
Formel: C₅H₇ClO₃
MW: 150,56 g/mol
Kokpunkt: 212…213 °C (760 mmHg)
Smältpunkt: 14…16 °C
Densitet: 1,28 g/cm³
Flampunkt: 107 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000938
CAS nummer: 32807-28-6
EINECS: 251-230-5
UN: 2922
ADR: 8,III

Order Now

Specifikation testresultat

Appearance Clear light yellow to light orange Liquid
Infrared spectrum Conforms
Refractive index 1.4560 to 1.4580 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR