7-Ethoxy-4-methylcoumarin

Leverantör: Thermo Scientific

SB00763EA
MAYBSB00763EAEA 0 SEK
MAYBSB00763EA
7-Ethoxy-4-methylcoumarin
7-Ethoxy-4-methylcoumarin
Formel: C₁₂H₁₂O₃
MW: 204,23 g/mol
MDL Number: MFCD00016971
CAS nummer: 87-05-8
EINECS: 201-721-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR