Bis(2-bromoethyl)ether 95%
Leverantör: Thermo Scientific
Fara
Synonyms: 1-Bromo-2-(2-bromoethoxy)ethane, 2-Bromoethyl ether
L11510.06 L11510.14
L11510.06EA 1250 SEK
L11510.06 L11510.14
Bis(2-bromoethyl)ether 95%
Bis(2-bromoethyl)ether
Purity: 96% CAS No.: 5414-19-7 Molecular Formula: BrCH2CH2OCH2CH2Br Formula Weight: 231.93 Flash Point: 85°(185°F) Refractive Index: 1.5140 Boiling Point: 92-93°/12mm Beilstein Reference: 1098335 UN No.: 2810 MDL No.: MFCD00039196
Formel: C₄H₈Br₂O
MW: 231,93 g/mol
Kokpunkt: 92…93 °C (12 mmHg)
Densitet: 1,845
Flampunkt: 85 °C (185 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00039196
CAS nummer: 5414-19-7
EINECS: 226-504-2
UN: 2810
ADR: 6.1,II