2-Imidazolidinone hemihydrate 99+%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Imidazolidone hemihydrate
122040025 122040250 122045000 122040100
ACRO122040025EA 3800 SEK
ACRO122040025 ACRO122040250 ACRO122045000 ACRO122040100
2-Imidazolidinone hemihydrate 99+%
2-Imidazolidinone hemihydrate
Formel: C₃H₆N₂O·0.5H₂O
MW: 95,10 g/mol (86,09 anhy)
Kokpunkt: >100 °C (760 mmHg)
Smältpunkt: 58 °C
Flampunkt: 265 °C
Storage Temperature: Omgivande
MDL Number: MFCD02662355
CAS nummer: 121325-67-5
EINECS: 204-436-4
Order Now
Specifikation testresultat
Appearance White to light yellow Adhering crystalline powder or crystals
Infrared spectrum Conforms
Water ≤13 %
GC ≥99.0 %
Elemental Analysis 5.1 to 5.6 % N