4-Iodobenzaldehyde ≥96.0%
Leverantör: TCI
Varning
I0612-1G I0612-5G
TCIAI0612-1GEA 685 SEK
TCIAI0612-1G TCIAI0612-5G
4-Iodobenzaldehyde ≥96.0%
4-Iodobenzaldehyde
Formel: C₇H₅IO
Kokpunkt: 265°C
MDL Number: MFCD00039576
CAS nummer: 15164-44-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f4034650d0260f2dc4225bb20ad761fab1a5039&vid=5f4034650d0260f2dc4225bb20ad761fab1a5039&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 96.0 %