1,7-Heptanediol ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 1,7-Dihydroxy heptane

H0028-25G H0028-5G
TCIAH0028-25GEA 1900 SEK
TCIAH0028-25G TCIAH0028-5G
1,7-Heptanediol ≥98.0%
1,7-Heptanediol
Formel: C₇H₁₆O₂
MW: 132,2 g/mol
Kokpunkt: 262°C
Smältpunkt: 17…18 °C
Densitet: 0,953 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002987
CAS nummer: 629-30-1
EINECS: 211-085-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4066912d71bd954a0084cda0652fae2b2ca4c960&vid=80036d3d4cf7142dcf8764f937b7c1ec8d77d82e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR