Ytterbium(III) chloride hexahydrate 99.99+% Spårmetallbas
Leverantör: Thermo Fisher Scientific
Varning
437180500 437180100
ACRO437180500EA 5600 SEK
ACRO437180500 ACRO437180100
Ytterbium(III) chloride hexahydrate 99.99+% Spårmetallbas
Ytterbium(III) chloride hexahydrate
Formel: Cl₃Yb·6H₂O
MW: 387.49 g/mol
Smältpunkt: 865 °C
Densitet: 2.57
Storage Temperature: Omgivande
MDL Number: MFCD00149885
CAS nummer: 10035-01-5
EINECS: 233-800-5
Order Now
Specifikation testresultat
Appearance White Powder or crystals
Total trace metal impurities ≤100 ppm