Natriumhexan-1-sulfonatmonohydrat, vattenfri 99+% för jonparkromatografi
Leverantör: Thermo Scientific
Varning
Synonyms: Sodium hexane-1-sulphonate, Sodium hexanesulphonate, Sodium 1-hexanesulphonate
396670250 396671000
ACRO396670250EA 7350 SEK
ACRO396670250 ACRO396671000
Natriumhexan-1-sulfonatmonohydrat, vattenfri 99+% för jonparkromatografi
Natriumhexan-1-sulfonatmonohydrat
Formel: C₆H₁₃O₃SNa
MW: 188,22 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007542
CAS nummer: 2832-45-3
EINECS: 220-601-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aebc3cb7a7cefac9ad5a7678d82a90d244b508f9&vid=f98fce9a53ea29283b11fb44162f26067930bbbe&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White Crystalline solid
Infrared spectrum Conforms
Loss on drying ≤2 % (150°C)
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.5 M soln.) Passes test
Suitability (HPLC gradient) Passes test