4,5-Difluoro-2-nitroaniline ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 3,4-Difluoro-6-nitroaniline
H55830.03 H55830.06
H55830.03EA 1250 SEK
H55830.03 H55830.06
4,5-Difluoro-2-nitroaniline ≥98%
4,5-Difluoro-2-nitroaniline
Purity: 98% CAS No.: 78056-39-0 Molecular Formula: C6H4F2N2O2 Formula Weight: 174.11 Melting Point: 107-108° Beilstein Reference: 2723242
Formel: C₆H₄F₂N₂O₂
MW: 174,11 g/mol
Smältpunkt: 107…108 °C
Storage Temperature: Omgivande
MDL Number: MFCD00010876
CAS nummer: 78056-39-0