Benzil ≥99.0%
Leverantör: TCI
Varning
Synonyms: 1,2-Diphenyl-1,2-ethanedione
B0050-25G B0050-500G
TCIAB0050-25GEA 215 SEK
TCIAB0050-25G TCIAB0050-500G
Benzil ≥99.0%
Benzil
Formel: (C₆H₅)C(O)C(O)(C₆H₅)
Kokpunkt: 348°C
Smältpunkt: 95 °C
Densitet: 1,23 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003080
CAS nummer: 134-81-6
EINECS: 205-157-0
Merck Index: 14,01078
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a18e91309875edb4d9a8c6319d11f7cc04690a7&vid=4a18e91309875edb4d9a8c6319d11f7cc04690a7&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 99.0 %
Melting point 94.0 to 97.0 deg-C
Solubility in hot EtOH almost transparency