N-Metyl-2-pyrrolidon (NMP) for peptide synthesis, Supelco®
Leverantör: Merck
Fara
M-Clarity™ quality level = MQ200
1.00574.2500 1.00574.4000
1.00574.2500EA 4780 SEK
1.00574.2500 1.00574.4000
N-Metyl-2-pyrrolidon (NMP) for peptide synthesis, Supelco®
N-Metyl-2-pyrrolidon (NMP)
Formel: C₅H₉NO
Kokpunkt: 202 °C (1013 hPa)
Smältpunkt: –24 °C
Densitet: 1,033 g/cm³ (20 °C)
Flampunkt: 91 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003193
CAS nummer: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM