SRK® Pharma Swab Rinse Set
Leverantör: Copan
958C 952C 9U004S
COPA958CEA 10450 SEK
COPA958C 710-1862 COPA9U004S
SRK® Pharma Swab Rinse Set
Svabbar Svabbar med sköljlösningar
Swab rinse kits (SRK®) for microbiological examination of surfaces.
  • Isotonic salt solution with neutralising substances
  • Deactivate disinfectants and sanitising agents
  • Rinse solution to moisten the sterile swab before sampling
Suitable for use during microbial detection or quantitative analysis.
Förpackning: Triple-protection wrapping. 20 sets per pack.