2,5-Dihydro-2,5-dimethoxyfuran (mixture of cis and trans isomers) ≥99%
Leverantör: Thermo Scientific
Fara
Synonyms: 2,5-Dimethoxy-2,5-dihydrofuran
B25028.0B B25028.22 B25028.30 B25028.36
B25028.0BEA 8625 SEK
B25028.0B B25028.22 B25028.30 B25028.36
2,5-Dihydro-2,5-dimethoxyfuran (mixture of cis and trans isomers) ≥99%
2,5-Dihydro-2,5-dimethoxyfuran (mixture of cis and trans isomers)
Purity: 99% CAS No.: 332-77-4 Molecular Formula: C6H10O3 Formula Weight: 130.14 Flash Point: 47°(116°F) Refractive Index: 1.4350 Melting Point: -40° Boiling Point: 160-162° Beilstein Reference: 113225 UN No.: 3271 MDL No.: MFCD00003220
Formel: C₆H₁₀O₃
MW: 130,14 g/mol
Kokpunkt: 160…162 °C
Smältpunkt: < –40 °C
Densitet: 1,072
Flampunkt: 47 °C (116 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00003220
CAS nummer: 332-77-4
EINECS: 206-367-5
UN: 3271
ADR: 3,III