2'-Deoxycytidine hydrochloride ≥99%
Leverantör: Thermo Scientific
Synonyms: 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one hydrochloride
L14153.03 L14153.MD
L14153.03EA 1400 SEK
L14153.03 L14153.MD
2'-Deoxycytidine hydrochloride ≥99%
2'-Deoxycytidine hydrochloride
Purity: 99% CAS No.: 3992-42-5 Molecular Formula: C9H13N3O4.HCl Formula Weight: 263.68 Odour: Odourless Melting Point: ca 170° dec. Beilstein Reference: 3576124 Optical Rotation: +57° (c=1 in water) MDL No.: MFCD00012840
Formel: C₉H₁₄ClN₃O₄
MW: 263,68 g/mol
Smältpunkt: ∼ 170 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00012840
CAS nummer: 3992-42-5
EINECS: 223-639-9