Adenosine-5'-monophosphoric acid (AMP), (max. 6% H₂O) 99%
Leverantör: Thermo Scientific
Synonyms: ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogen phosphate, 5'-Adenylic acid, Adenosine 5'-monophosphate, Adenosine monophosphate, 5'-AMP
L14051.03 L14051.06
L14051.03EA 485 SEK
L14051.03 L14051.06
Adenosine-5'-monophosphoric acid (AMP), (max. 6% H₂O) 99%
Adenosine-5'-monophosphoric acid (AMP)
a useful ligand determinant that facilitate the binding of APS reductase inhibitors and activates adenosine receptor agonists.
5'-AMP is an activator of a class of protein kinases known as AMP-activated protein kinase, an activator of phosphorylase b in the polymerization reaction for the synthesis of branched polysaccharides, a reagent in the synthesis of adenosine-5?-phosphoimidazolide, an inhibitor of endogenous NMN adenylyltransferase (NMNAT) activity that converts NMN to NAD+.
Formel: C₁₀H₁₄N₅O₇P
MW: 347,22 g/mol
Smältpunkt: ∼ 190 °C (dec.)
Storage Temperature: Kylskåp
MDL Number: MFCD00149360
CAS nummer: 61-19-8
EINECS: 200-500-0
Merck Index: 14,158