Adenosine 5'-diphosphate disodium salt (ADP disodium salt), (max. 7% H₂O) 96%
Leverantör: Thermo Fisher Scientific
Synonyms: Adenosine 5'-Diphosphate Disodium Salt, Sodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl hydrogen diphosphate
L14029.03 L14029.MD
L14029.03EA 1300 SEK
L14029.03 L14029.MD
Adenosine 5'-diphosphate disodium salt (ADP disodium salt), (max. 7% H₂O) 96%
Adenosine 5'-diphosphate disodium salt (ADP disodium salt)
Purity: 97% CAS No.: 16178-48-6 Molecular Formula: C10H13N5Na2O10P2 Formula Weight: 471.17 Storage Temperature: -30°C to -10°C MDL No.: MFCD00066635
Formel: C₁₀H₁₃N₅Na₂O₁₀P₂
MW: 471.167 g/mol
Storage Temperature: Frys
MDL Number: MFCD00066635
CAS nummer: 16178-48-6
EINECS: 240-314-7