Guanosine 5'-triphosphate disodium salt hydrate 90% för biokemi
Leverantör: Thermo Scientific
Synonyms: GTP disodium salt hydrate
226250010 226250500 226252500
ACRO226250010EA 11200 SEK
ACRO226250010 ACRO226250500 ACRO226252500
Guanosine 5'-triphosphate disodium salt hydrate 90% för biokemi
Guanosine 5'-triphosphate disodium salt hydrate
Formel: C₁₀H₁₄N₅Na₂O₁₄P₃·H₂O
MW: 567,147 g/mol (anhy)
Smältpunkt: 180 °C (dec.)
Storage Temperature: Frys
MDL Number: MFCD03410297
CAS nummer: 56001-37-7
EINECS: 259-940-7
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤12 % (K.F.)
HPLC ≥88 %
UV spectrophotometry ≥88 % (lambda max. 252 nm, pH 7)
Ratio of Absorbances 0.63 to 0.69 (A280/A260)
Related substances ≤4 % (typically) Guanosine diphosphate (HPLC)
Sodium (Na) 7 to 9.5 % (flame photom.)