Sudan red 7B (fat red bluish, solvent red 19, fat red 7B)
Leverantör: Thermo Scientific
Synonyms: N-Ethyl-1-((4-(phenyldiazenyl)phenyl)diazenyl)naphthalen-2-amine
191630050 191630500
ACRO191630050EA 1050 SEK
ACRO191630050 ACRO191630500
Sudan red 7B (fat red bluish, solvent red 19, fat red 7B)
Sudan red 7B (fat red bluish, solvent red 19, fat red 7B)
Formel: C₂₄H₂₁N₅
MW: 379,4570 g/mol
Smältpunkt: 130 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00003904
CAS nummer: 6368-72-5
EINECS: 228-862-5
Order Now
Specifikation testresultat
Appearance Bordeaux Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies