Isethionic acid sodium salt (sodium 2-hydroxyethanesulphonate) ≥98%
Leverantör: Thermo Scientific
Synonyms: Sodium isethionate, Sodium hydroxyethyl sulphonate, 2-Hydroxyethanesulphonic acid sodium salt
A12054.0B A12054.0I
A12054.0BEA 1900 SEK
A12054.0B A12054.0I
Isethionic acid sodium salt (sodium 2-hydroxyethanesulphonate) ≥98%
Isethionic acid sodium salt (sodium 2-hydroxyethanesulphonate)
Purity: 98% CAS No.: 1562-00-1 Molecular Formula: HO(CH2)2SO3Na Formula Weight: 148.11 Melting Point: 191-194° Beilstein Reference: 3633992
Formel: C₂H₅NaO₄S
MW: 148,11 g/mol
Smältpunkt: 191…194 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007534
CAS nummer: 1562-00-1
EINECS: 216-343-6