N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA) ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: N-(tert-Butyldimethylsilyl)-2,2,2-trifluoro-N-methylacetamide, MBDSTFA
B23760.03 B23760.06 B23760.14
B23760.03EA 925 SEK
B23760.03 B23760.06 B23760.14
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA) ≥97%
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA)
Purity: 97% CAS No.: 77377-52-7 Molecular Formula: C9H18F3NOSi Formula Weight: 241.33 Flash Point: 52°(125°F) Refractive Index: 1.4020 Boiling Point: 168-170° Beilstein Reference: 3606546 UN No.: 1993 MDL No.: MFCD00009671
Formel: C₉H₁₈F₃NOSi
MW: 241,33 g/mol
Kokpunkt: 168…170 °C
Densitet: 1,023
Flampunkt: 52 °C (125 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009671
CAS nummer: 77377-52-7
ADR: 3,III