N-Methyl-bis-trifluoroacetamide 98%
Leverantör: Thermo Scientific
Fara
Synonyms: 2,2,2-Trifluoro-N-methyl-N-(2,2,2-trifluoroacetyl)acetamide, MBTFA
210070050 210070250
ACRO210070050EA 3700 SEK
ACRO210070050 ACRO210070250
N-Methyl-bis-trifluoroacetamide 98%
N-Methylbistrifluoroacetamide
Formel: C₅H₃F₆NO₂
MW: 223,07 g/mol
Kokpunkt: 120…122 °C (760 mmHg)
Densitet: 1,547 g/cm³
Flampunkt: 45 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000412
CAS nummer: 685-27-8
EINECS: 211-680-5
UN: 2920
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3450 to 1.3470 (20°C, 589 nm)
GC ≥97.5 %