Potassium xanthogenate (potassium-O-ethyl dithiocarbonate) 97+%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Potassium-O-ethyldithiocarbonate, Potassium ethyl xanthate, Ethylxanthic acid potassium salt
410500250 410501000 410505000
ACRO410500250EA 505 SEK
ACRO410500250 ACRO410501000 ACRO410505000
Potassium xanthogenate (potassium-O-ethyl dithiocarbonate) 97+%
Potassium xanthogenate (potassium-O-ethyl dithiocarbonate)
Formel: C₃H₅KOS₂
MW: 160.30 g/mol
Smältpunkt: 209…214 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00004931
CAS nummer: 140-89-6
EINECS: 205-439-3
Order Now
Specifikation testresultat
Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 %
Titration with HClO4 ≥97.0 % (On dry substance)