o-Cresolphthalein complexone indicator for complexometry, Fluka™
Leverantör: Honeywell Chemicals
Synonyms: o-Cresolphthalein Complexone, Phthalein purple, Cresol phthalexone, Xylenolphthalein-bis(iminodiacetic acid), Phthalein complexone, Phthalate complexing agent [Chelation indicator], Metal phthaleine, o-Cresolphthalexon
64000-25G 64000-5G
64000-25GEA 6500 SEK
64000-25G 64000-5G
o-Cresolphthalein complexone indicator for complexometry, Fluka™
o-Cresolphthalein complexone
Formel: C₃₂H₃₂N₂O₁₂
MW: 636.62 g/mol
Smältpunkt: 181…185 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00005911
CAS nummer: 2411-89-4
EINECS: 219-318-8
Order Now
Specifikation testresultat
Ignition Residue ~1%
Appearance of the substance complying
Identity (LC-MS) complying
Indicator solution complying
Water (Karl Fischer) Max. 10 %
Residue on ignition Max. 2 %
Suitability for Complexometry complying