o-Cresolphthalein complexone Reag. Ph. Eur. indicator for metal titration, Supelco®
Leverantör: Merck
Synonyms: o-Cresolphthalein Complexone, Phthalein purple, Cresol phthalexone, Xylenolphthalein-bis(iminodiacetic acid), Phthalein complexone, Phthalate complexing agent [Chelation indicator], Metal phthaleine, o-Cresolphthalexon
M-Clarity™ quality level = MQ200
1.07297.0001 1.07297.0010
1.07297.0001EA 4300 SEK
1.07297.0001 1.07297.0010
o-Cresolphthalein complexone Reag. Ph. Eur. indicator for metal titration, Supelco®
o-Cresolphthalein complexone
Formel: C₃₂H₃₂N₂O₁₂
MW: 636.62 g/mol
Smältpunkt: 186 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005911
CAS nummer: 2411-89-4
EINECS: 219-318-8
Order Now
Specifikation testresultat
Identity (UV/VIS-Spectrum) passes test
Absorption maximum λmax. (Sodium hydroxide solution 0.1 mol/l) 577 - 580 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l; sodium hydroxide solution 0.1 mol/l; calc. on anhydrous substance) ≥ 750
Water (according to Karl Fischer) ≤ 10 %
Suitability as indicator (for metal titration) passes test