1-Naphthyl phosphate monosodium salt monohydrate ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Naphthylphosphoric Acid Monosodium Salt Monohydrate, Monosodium 1-Naphthyl Phosphate Monohydrate, Sodium 1-Naphthyl Hydrogen Phosphate Monohydrate, Phosphoric Acid 1-Naphthyl Ester Monosodium Salt Monohydrate
A18254.06 A18254.14 A18254.22
A18254.06EA 2000 SEK
A18254.06 A18254.14 A18254.22
1-Naphthyl phosphate monosodium salt monohydrate ≥98%
1-Naphthyl phosphate monosodium salt monohydrate
Purity: 98+% CAS No.: 81012-89-7 Molecular Formula: C10H8NaO4P.H2O Formula Weight: 264.16 (246.14anhy) Storage Temperature: 2°C to 8°C Melting Point: 186-191° MDL No.: MFCD00150615
Formel: C₁₀H₈NaO₄P·1H₂O
MW: 264,15 g/mol
Smältpunkt: 186…191 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00150615
CAS nummer: 81012-89-7
EINECS: 220-171-7