FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) 90%, ren
Leverantör: Thermo Scientific
Fara
Synonyms: 5-isothiocyanatofluorescein, Fluorescein 5(6)-isothiocyanate, Fluorescein Isothiocyanate, Isomer I
119250010 119252500 119250050
ACRO119250010EA 4525 SEK
ACRO119250010 ACRO119252500 ACRO119250050
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) 90%, ren
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I)
Formel: C₂₁H₁₁NO₅S
MW: 389,381 g/mol
Smältpunkt: >360 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00005063
CAS nummer: 3326-32-7
EINECS: 222-042-0
Order Now
Specifikation testresultat
Appearance Orange Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %